missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-3 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
Marca: R&D Systems MAB403-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IL-3 Monoclonal specifically detects IL-3 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Especificaciones
| IL-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3562 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, ELISA | |
| MP28F8 | |
| Western Blot 1 ug/mL, Neutralization 0.05-0.15 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P01586 | |
| IL3 | |
| COS-7 African green monkey SV40 transformed kidney fibroblast-like cell line-derived recombinant mouse IL-3 Accession # P01586 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) IL-3 or recombinant rat IL-3 is observed. Does not neutralize the biological activity of rhIL-3. | |
| Mouse | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido