missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IL-18 Recombinant Protein

Código de producto. 10252533
Change view
Click to view available options
Cantidad:
25 μg
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
10252533 25 μg cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10252533 Proveedor Gibco™ N.º de proveedor PMC0184

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Reconstitution: We recommend that the vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute the contents of the vial with 250 μL ice cold, sterile, distilled water on ice. Storage: Store this lyophilized preparation at -20°C, preferably desiccated. Upon reconstitution, the protein may be stored at 2°C to 8°C for up to one week. For long term storage of the reconstituted protein, apportion into working aliquots and store at -80°C. Avoid repeated freeze/thaw cycles.

The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P70380
Para utilizar con (aplicación) Bioactivity
Formulación PBS with 0.1% BSA, 1% sucrose and no preservative
ID de gen (Entrez) 16173
Peso molecular 18 kDa
Nombre Mouse IL-18
Método de purificación Purified
Cantidad 25 μg
Requisitos de almacenamiento -20°C
Estado normativo RUO
Concentración de endotoxinas <0.1 ng/μg
Alias de gen cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320
Nombre común IL-18
Símbolo de gen IL18
Actividad biológica Determined by the dose-dependent induction of IFN-gamma production in mouse IL-18 receptor transfected KG-1 cells.
Tipo de producto Protein
Conjugado Unconjugated
Especie Mouse
Recombinante Recombinant
Secuencia Mouse IL-18, amino acids 36-192 of the mature sequence
Contenido y almacenamiento -20°C
Actividad Determined by the dose-dependent induction of IFN-gamma production in mouse IL-18 receptor transfected KG-1 cells.
Nivel de endotoxinas <0.1 ng/μg
Condiciones de envío Approved for shipment on Wet or Dry Ice
Sistema de expresión E. coli
Familia de proteínas Cytokines & Receptors
Forma de proteína Recombinant, Ligand
Formulario Lyophilized
Subtipo de proteína Interleukins
Categoría de investigación Immunology, Neurobiology, Inflammation, Angiogenesis, Signal Transduction
Línea de productos Gibco
Grado de pureza o calidad >90% by SDS-PAGE
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.