missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse Granzyme B Recombinant Protein, PeproTech®

Código de producto. 17821653
Change view
Click to view available options
Cantidad:
10 x 100 μg
100 μg
10 μg
2,5 x 100 μg
2 μg
5 x 100 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
2 microgramos
250 microgramos
500 microgramos
paquete de 5
Código de producto. Cantidad unitSize
17821653 2 μg 2 microgramos
17830503 100 μg 100 microgramos
17841543 10 μg 10 microgramos
30087491 5 x 100 μg paquete de 5
17830983 10 x 100 μg 1 miligramo
17890193 2,5 x 100 μg 250 microgramos
17870023 5 x 100 μg 500 microgramos
7 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17821653

Marca: Gibco™ 140032UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

140-03-1MG will be provided as 10 x 100 μg (140-03-100UG). 140-03-250UG will be provided as multiple 50 μg (140-03-50UG) and 100 μg (140-03-100UG) vials. 140-03-500UG will be provided as 5 x 100 μg (140-03-100UG). Recombinant Murine Granzyme B is a glycosylated 227 amino acid protein, comprising the mature active portion of the murine Granzyme B precursor. The apparent molecular weight is 28.9 kDa by mass spectrometry. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Granzyme B is a member of the granzyme serine protease family, and is found in the granules of cytotoxic T cells and NK cells. Granzyme B has been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved in this process has been identified as mannose 6-phosphate receptor which functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis. Granzyme B enters target cells to cleave caspase-3 and initiate the caspase cascade leading to DNA fragmentation and apoptosis. Granzyme B can also act through a mitochondrial apoptosis pathway by cleaving the Bid protein. Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P04187
Para utilizar con (aplicación) Control, Bioactivity
Formulación Protein with no preservative
ID de gen (Entrez) 14939
Peso molecular 28.9 kDa
Nombre Mouse Granzyme B
Método de purificación Purified
Cantidad 2 μg
Fuente Baculovirus
Requisitos de almacenamiento -20°C
Estado normativo RUO
Concentración de endotoxinas <1 EU/μg
Alias de gen AI553453; C11; Cathepsin G-like 1; CCP1; CCP-1/C11; CCPI; CGL1; CGL-1; CSPB; CSP-B; Ctla1; Ctla-1; CTSGL1; cytotoxic cell protease 1; cytotoxic serine protease B; Cytotoxic T lymphocyte associated serine esterase 1; cytotoxic T-lymphocyte proteinase 2; cytotoxic T-lymphocyte-associated serine esterase 1; fragmentin; fragmentin 2; fragmentin-2; GLP I; GLP III; GLP-1; granzyme 2; granzyme B; granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1); granzyme B(G,H); Granzyme-2; GranzymeB; granzyme-like protein 1; granzyme-like protein I; GRB; GZB; Gzmb; HLP; human lymphocyte protein; Human lymphocyte protein (Hlp); Lymphocyte protease; Natural killer cell protease 1; OTTHUMP00000028189; RNKP-1; SECT; T-cell serine protease 1-3E
Nombre común Granzyme B
Símbolo de gen GZMB
Actividad biológica Determined by its ability to cleave a synthetic chromogentic Granzyme B substrate. The expected specific activity, when using the Ac-IEPD-pNA substrate at 25°C, is greater than 750 nM/min / ug of enzyme.
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Secuencia IIGGHEVKPH SRPYMALLSI KDQQPEAICG GFLIREDFVL TAAHCEGSII NVTLGAHNIK EQEKTQQVIP MVKCIPHPDY NPKTFSNDIM LLKLKSKAKR TRAVRPLNLP RRNVNVKPGD VCYVAGWGRM APMGKYSNTL QEVELTVQKD RECESYFKNR YNKTNQICAG DPKTKRASFR GDSGGPLVCK KVAAGIVSYG YKDGSPPRAF TKVSSFLSWI KKTMKSS
Contenido y almacenamiento -20°C
Actividad Determined by its ability to cleave a synthetic chromogentic Granzyme B substrate. The expected specific activity, when using the Ac-IEPD-pNA substrate at 25°C, is greater than 750 nM/min per ug of enzyme.
Sistema de expresión Baculovirus
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.