missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse CXCL9 (MIG) Recombinant Protein, PeproTech®
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
250 μg
500 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
5 microgramos
500 microgramos
Descripción
Recombinant Murine MIG is a 12.2 kDa protein containing 105 amino acid residues, including the four highly conserved cysteine residues present in CXC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
MIG, also known as CXCL9, is a T-cell chemoattractant inducible by gamma interferon that is a member of the CXC chemokine family of cytokines.This gene is mapped to 4q21. It is noted that, although the best-described activities of the chemokines are as chemotactic factors, chemokines also have an effect on T-cell activation, angiogenesis, and HIV infection. While most CXC chemokines are chemotactic for neutrophils, MIG and INP10 are unusual and similar in being CXC chemokines that are chemotactic for lymphocytes and inactive in neutrophils.
Especificaciones
Especificaciones
| Número de acceso | P18340 |
| Para utilizar con (aplicación) | Functional Assay |
| Formulación | protein with no preservative |
| ID de gen (Entrez) | 17329 |
| Peso molecular | 12.2 kDa |
| Nombre | Mouse CXCL9 (MIG) |
| Cantidad | 250 μg |
| Fuente | E. coli |
| Estado normativo | RUO |
| Concentración de endotoxinas | <1 EU/ μg |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido