missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse CXCL16 Recombinant Protein, PeproTech®
Click to view available options
Cantidad:
100 μg
1 mg
250 μg
25 μg
500 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
25 microgramos
250 microgramos
5 microgramos
500 microgramos
Descripción
Recombinant Murine CXCL16 is a 9.9 kDa protein containing 88 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
CXCL16 is a member of the CXC chemokine family and signals through the CXCR6 receptor. CXCL16 may play a role in attracting lymphocyte subsets during inflammation and may facilitate certain immune responses. The chemokine domain of CXCL16 contains six cysteine residues, including the four highly conserved cysteine residues characteristic of CXC chemokines. The CXCL16 gene codes for a 273 amino acid polypeptide, which includes a 29 amino acid cytoplasmic domain and transmembrane sequence containing approximately 20 amino acids. The extracellular portion of CXCL16 contains a chemokines domain and an extended C-terminal mucin-like stalk sequence. The extracellular domain contains 89 amino acid residues (86 a.a. residues for the murine homolog).
Especificaciones
Especificaciones
| Número de acceso | Q8BSU2 |
| Para utilizar con (aplicación) | ELISA, Functional Assay, Western Blot |
| Formulación | protein with no preservative |
| ID de gen (Entrez) | 66102 |
| Peso molecular | 9.9 kDa |
| Nombre | Mouse CXCL16 |
| Cantidad | 1 mg |
| Fuente | E. coli |
| Estado normativo | RUO |
| Concentración de endotoxinas | <1 EU/ μg |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido