missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Complement Component C5a Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Marca: R&D Systems MAB21501-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Complement Component C5a Monoclonal specifically detects Complement Component C5a in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Especificaciones
| Complement Component C5a | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C5, C5A, HE | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 727 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Neutralization, ELISA | |
| 295108 | |
| Western Blot 1 ug/mL, Neutralization 0.1-0.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P06684 | |
| Hc | |
| E. coli-derived recombinant mouse Complement Component C5a Asn679-Arg755 Accession # P06684 | |
| 25 μg | |
| Primary | |
| Detects mouse Complement Component C5a in Western blots. In Western blots, this antibody does not cross-react with recombinant mouse Complement Component C3d, recombinant human (rh) Complement Component C3a, or rhComplement Component C5a. | |
| Mouse | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido