missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Complement Component C3aR Antibody, R&D Systems™

Rat Monoclonal Antibody
Marca: R&D Systems MAB10417-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Complement Component C3aR Monoclonal specifically detects Complement Component C3aR in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Especificaciones
| Complement Component C3aR | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09 | |
| Chinese hamster ovary cell line CHO-derived transfected with mouse C3aR, Accession # O09047 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 620613 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| O09047 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 719 | |
| Mouse | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido