missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL12/MCP-5 Antibody, R&D Systems™

Rat Monoclonal Antibody
Marca: R&D Systems MAB428-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CCL12/MCP-5 Monoclonal specifically detects CCL12/MCP-5 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Especificaciones
| CCL12/MCP-5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 12, chemokine (C-C motif) ligand 12, MCP5, MCP-5, Scya12 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 20293 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Neutralization, ELISA | |
| 63205 | |
| Western Blot 1 ug/mL, Neutralization 0.5-2.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q62401.1 | |
| Ccl12 | |
| E. coli-derived recombinant mouse CCL12/MCP-5 Gly23-Gly104 Accession # Q62401.1 | |
| 100 μg | |
| Primary | |
| Detects mouse CCL12/MCP-5 in ELISAs and Western blots. | |
| Mouse | |
| Supernatant |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido