Learn More
Mouse anti-Human IgG1 Fc Secondary Antibody (2C11cc), HRP, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
IgG1 Fc Monoclonal antibody specifically detects IgG1 Fc in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunodiffusion
Especificaciones
Especificaciones
| Antígeno | IgG1 Fc |
| Aplicaciones | ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunodiffusion |
| Clasificación | Monoclonal |
| Clon | 2C11cc |
| Conjugado | HRP |
| Dilución | ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunodiffusion |
| Formulación | PBS |
| Especie del huésped | Mouse |
| Inmunógeno | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG from human serum. |
| Método de purificación | Protein A purified |
| Mostrar más |