Learn More
CD42b Mouse anti-Human, Brilliant Violet 421, Clone: HIP1, BD Optibuild™
Mouse Monoclonal Antibody
Marca: BD Biosciences 740075
Descripción
The HIP1 monoclonal antibody specifically binds to CD42b. CD42b is also known as the Platelet glycoprotein Ib alpha chain that is encoded by the GP1BA gene. CD42b is disulfide bonded to CD42c to form a 170 kDa heterodimer, GPIb. GPIb forms a noncovalent complex with CD42a and CD42d (CD42 complex) that is expressed on platelets and megakaryocytes. The CD42 complex serves as the von Willebrand Factor (vWF) surface receptor involved in the adhesion of platelets to the subendothelium of damaged vascular walls. HIP1 inhibits the ristocetin-dependent binding of vWF to platelets and partially inhibits collagen-induced aggregation.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Especificaciones
CD42b | |
Monoclonal | |
0.2mg/mL | |
GP1BA; GPIbalpha; GP-Ib alpha; GPIb-alpha; CD42b-alpha; Glycocalicin; BSS | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
HIP1 | |
Brilliant Violet 421 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
For Research Use Only