missing translation for 'onlineSavingsMsg'
Learn More

CD144 Mouse anti-Human, Brilliant Violet 421, Clone: 55-7H1, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 565670

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15894449

  • 510.00€ / cada uno
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

The 55-7H1 antibody monoclonal antibody specifically binds to a calcium-independent epitope on Cadherin-5, a member of the cadherin family of calcium-dependent adhesion molecules. Cadherin-5 is also known as CD144 or VE-Cadherin. CD144 is expressed on endothelial cells in vivo and in vitro. It may play a role in the organization of lateral endothelial junctions and in the control of permeability properties of vascular endothelium.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

Especificaciones

Especificaciones

CD144
Monoclonal
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Human
IgG1 κ
Flow Cytometry
55-7H1
VE-cadherin; Cadherin-5; CDH5; Vascular endothelial cadherin
Mouse
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.