Learn More
Novus Biologicals™ MORN1 Recombinant Protein Antigen
Código de producto. 18027157 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μl
Tamaño de la unidad:
100 microlitros
18027157 100 μl 100 microlitros
1 options
Código de producto. 18027157
Marca: Novus Biologicals™ NBP194013PEP
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Antibody Competition
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human MORN1
The MORN1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Specifica
Gene ID (Entrez) 79906
Especie Human
Pureza >80% by SDS-PAGE and Coomassie blue staining
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Para utilizar con (aplicación) Antibody Competition
Símbolo de gen MORN1
Tipo de etiqueta Unconjugated
Peso molecular 32 kDa
Cantidad 100 μl
Estado normativo RUO
Fuente E. coli
Reactividad específica This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54930. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Tampón de almacenamiento PBS and 1M Urea, pH 7.4.
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.