Learn More
Monomethyl auristatin F (MMAF) Antibody (01), Novus Biologicals™
Código de producto. 30113564 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
20 μg
Tamaño de la unidad:
100 microgramos
20 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30113564 20 μg 20 microgramos
30116222 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30113564 Proveedor Novus Biologicals N.º de proveedor NBP33197520ug
Mouse Monoclonal Antibody
Monomethyl auristatin F (MMAF) Monoclonal antibody specifically detects Monomethyl auristatin F (MMAF) in Nonspecies specific samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Monomethyl auristatin F (MMAF)
Aplicaciones ELISA
Clasificación Monoclonal
Clon 01
Conjugado Unconjugated
Dilución ELISA 1:1000-1:2000
Formulación 0.2 um filtered solution in PBS
Alias de gen 745017-94-1, mafodotin, MMAF (GMP)
Especie del huésped Mouse
Inmunógeno This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with Monomethyl auristatin F (MMAF) conjugated to BSA
Método de purificación Protein A purified
Cantidad 20 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Non-species specific
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG1