Learn More
Monomethyl auristatin E (MMAE) Antibody (08), Novus Biologicals™
Código de producto. 30116704 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
20 μg
Tamaño de la unidad:
100 microgramos
20 microgramos
30116704 100 μg 100 microgramos
30115289 20 μg 20 microgramos
2 options
Código de producto. 30116704
Marca: Novus Biologicals NBP331974100ug
Mouse Monoclonal Antibody
Monomethyl auristatin E (MMAE) Monoclonal antibody specifically detects Monomethyl auristatin E (MMAE) in Nonspecies specific samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Monomethyl auristatin E (MMAE)
Aplicaciones ELISA
Clasificación Monoclonal
Clon 08
Conjugado Unconjugated
Dilución ELISA 1:1000-1:2000
Formulación 0.2 um filtered solution in PBS
Alias de gen 474645-27-7, Monomethyl auristatin E (GMP), vedotin
Especie del huésped Mouse
Inmunógeno This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with Monomethyl auristatin E (MMAE) conjugated to BSA
Método de purificación Protein A purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Non-species specific
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG1