Learn More
250MG 5'-Aminohexyl Linker Phosphoramidite
Marca: Thermo Scientific 27-0035-04
250MG 5'-Aminohexyl Linker Phosphoramidite
Detalles adicionales : Peso : 0.00020kg
Descripción
Thermo Scientific™ Modifying Phosphoramidites are for the addition of a primary amine to the 5' position of an oligonucleotide or to incorporate an amine within the sequence and Phosphorylating Phosphoramidites are used for addition of a phosphate group to the 3' or 5' position of an oligonucleotide.
Process and Supply Chain Control
- A controlled raw material supply chain and established vendor partnerships ensure consistency and security of supply
- Processes are strictly controlled and monitored from material procurement through final QC and product release under ISO9001 standards, providing batch-to-batch reproducibility
- All modifiers are supplied with lot-specific quality control data including HPLC and phosphorous NMR analyses
Amino Modifiers:
- The amino modification provides a site for attachment of fluorescent labels such as biotin or NuLight™ esters
- Oligos with an amino modification can also be attached to surfaces in DNA chip applications
- The 5' amino modifier amidites are supplied with MMT or TFA protection
- The MMT modifier allows for trityl-on purification of the resulting oligonucleotide
- The TFA group is removed during ammonia deprotection
Phosphorylation:
- The phosphorylating phosphoramidte is for the addition of a phosphate to the 5' or 3' position of the oligonucleotide during standard synthesis protocols
Flexible Packaging:
- Common pack sizes fit most commercially available synthesizers and are in stock for immediate delivery
- Custom and bulk packaging, including multikilogram lots, are available