Learn More
MMR/CD206/Mannose Receptor Antibody, Novus Biologicals™
Código de producto. 18039625 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
25 μL
0.1 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18039625 0.1 mL 0.10 ml
18465212 25 μL 25 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18039625 Proveedor Novus Biologicals N.º de proveedor NBP190020
Rabbit Polyclonal Antibody has been used in 8 publications
MMR/CD206/Mannose Receptor Polyclonal specifically detects MMR/CD206/Mannose Receptor in Human, Mouse, Porcine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno MMR/CD206/Mannose Receptor
Aplicaciones Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 0.04-0.4 ug/ml, Flow Cytometry Validated for Flow from a verified customer review., Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence Reported in scientific literature (PMID 28620274)., Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulación PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Alias de gen CD206, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen
Símbolos de los genes MRC1
Especie del huésped Rabbit
Inmunógeno This antibody was developed against Recombinant Protein corresponding to amino acids:NEDHKRCVDAVSPSAVQTAACNQDAESQKFRWVSESQIMSVAFKLCLGVPSKTDWVAITLYACDSKSEFQKWECKNDTLLGIKGEDLFFNYGNRQEKNIMLYKGSGLWSRWKIYG
Método de purificación Affinity Purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4360
Especificidad de la prueba Specificity of human MMR/CD206/Mannose Receptor antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human, Mouse, Pig
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only