missing translation for 'onlineSavingsMsg'
Learn More

MMR/CD206/Mannose Receptor Antibody (CL0387) [PE], Novus Biologicals™

Código de producto. 30292897 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30292897 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Product Code. 30292897 Supplier Novus Biologicals Supplier No. NBP252927PE

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMR/CD206/Mannose Receptor Monoclonal antibody specifically detects MMR/CD206/Mannose Receptor in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno MMR/CD206/Mannose Receptor
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CL0387
Conjugado PE
Formulación PBS
Alias de gen CD206, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from P22897, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Immunology, Innate Immunity, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 4360
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.