missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
MMP-3 Monoclonal antibody specifically detects MMP-3 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array
Especificaciones
Especificaciones
| Antígeno | MMP-3 |
| Aplicaciones | ELISA, Immunohistochemistry (Paraffin), Protein Array |
| Clasificación | Monoclonal |
| Clon | MMP3/2806 |
| Conjugado | Alexa Fluor 350 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1 |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant full-length human MMP-3 protein (Uniprot: P08254 ) |
| Método de purificación | Protein A purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?