Learn More
MLKL Mouse anti-Human, Mouse, Clone: 3D4C6, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66675-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.Especificaciones
| MLKL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8NB16, Q9D2Y4 | |
| Mlkl | |
| MLKL Fusion Protein Ag15188 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 3D4C6 | |
| Unconjugated | |
| MLKL | |
| FLJ34389, MLKL | |
| Mouse | |
| Protein G | |
| RUO | |
| 197259, 74568 | |
| -20°C | |
| Liquid |