Description
The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
Specifications
Specifications
| Antígeno | MLKL |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Monoclonal |
| Clon | 3D4C6 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | MLKL |
| N.º de referencia del gen | Q8NB16, Q9D2Y4 |
| Alias de gen | FLJ34389, MLKL |
| Show More |