Learn More
Berkshire™ MicroPolx™ 2750 Microdenier Wiper
Código de producto. p-200022127 Tienda Berkshire Productos
Click to view available options
Dimensiones:
23 x 23 cm (9 x 9 in)
Tamaño de la unidad:
paquete de 100
Código de producto. 17826997
Marca: Berkshire™ MPX2750.0909.10
Solo quedan null
Añadir a la cesta
Edge
Añadir a la cesta
MicroPolx™ 2750 is a cleanroom laundered polyester/nylon microfiber knitted wiper.
  • Ultrafine microfiber yarn increases surface area and provides superior wiping efficiency
  • Wedge shaped fiber cross section lifts, entraps and removes particulates from critical surfaces
  • Cleans efficiently with less solvent usage than traditional wipers
  • Removes oily soils with little or no solvent required
  • Soft hand and non-abrasive ultrasonic sealed edge are excellent for sensitive or scratch prone surfaces
  • Continuous filament yarns and tight knit construction provide excellent abrasion resistance and low fiber shedding
  • Chemically compatible with IPA and other commonly used solvents
Especificaciones
Dimensiones 23 x 23 cm (9 x 9 in)
Tipo de producto Flat Head Mop Cover
Longitud (imperial) 9 in
Esterilidad No estéril
Longitud (métrico) 23 cm
Anchura (métrico) 23 cm
Anchura (imperial) 9 in.
Desechables Un solo uso
Material Poliéster, Synthetic Microfiber