Learn More
Micronova™ MicroLoop™ Mop Heads
Tienda Micronova ProductosDescripción
• ISO Class 5 to 8 (Class 100 to 100,000)
• Durable looped construction
Sturdy and functional looped mop heads intended for use in service areas outside of cleanrooms.
ML-1 MicroLoop PVA mop head is ideal for clean-build projects, post-construction, and cleanroom support. ML-9 MicroLoop Polyester mop head is an economical choice for hazardous materials cleanup and general spill control.
Información de órdenes
Fíjelo a un mango de acero inoxidable (se vende por separado) con el adaptador de rosca 19-035-362B.
Compliance
Both mop heads are rated for ISO Class 5 to 8 (Class 100 to 100,000) environments.
Especificaciones
Especificaciones
| Tipo de producto | MicroLoop Mop |
| Esterilizables en autoclave | Esterilizable en autoclave |
| Para utilizar con (aplicación) | Ideal for clean build projects, post construction, cleanroom support |
| Material | Poliéster |