missing translation for 'onlineSavingsMsg'
Learn More

mGluR2 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Código de producto. 18539611 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
18539611 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 18539611

Brand: Novus Biologicals NBP297220AF594

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

mGluR2 Polyclonal antibody specifically detects mGluR2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno mGluR2
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Alexa Fluor 594
Formulación 50 mM Sodium Borate
Alias de gen GLUR2, glutamate metabotropic receptor 2, glutamate receptor, metabotropic 2, GPRC1Bmetabotropic glutamate receptor 2, mGlu2, mGluR2, MGLUR2glutamate receptor homolog
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with E. coli-derived Human mGluR2 fragment.
Método de purificación Antigen and protein A Affinity-purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Tumor Suppressors, Vision
Primario o secundario Primary
ID de gen (Entrez) 2912
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.