missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
mGluR1 Monoclonal antibody specifically detects mGluR1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
Specifications
| Antígeno | mGluR1 |
| Aplicaciones | Flow Cytometry, CyTOF |
| Clasificación | Monoclonal |
| Clon | 511601 |
| Conjugado | DyLight 350 |
| Dilución | Flow Cytometry, CyTOF-ready |
| Formulación | 50mM Sodium Borate |
| Alias de gen | glutamate receptor, metabotropic 1, GRM1A, metabotropic glutamate receptor 1, mGluR1, MGLUR1A, MGLUR1GPRC1AmGlu1 |
| Especie del huésped | Mouse |
| Inmunógeno | Chinese hamster ovary cell line CHO-derived recombinant human mGluR1a, Met1-Ser522, Accession # Q13255 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?