missing translation for 'onlineSavingsMsg'
Learn More

MGA Antibody, Alexa Fluor™ 594, Novus Biologicals™

Código de producto. 18007608 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18007608 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18007608 Proveedor Novus Biologicals N.º de proveedor NBP298097AF594

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MGA Polyclonal specifically detects MGA in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno MGA
Aplicaciones Immunofluorescence
Clasificación Polyclonal
Conjugado Alexa Fluor 594
Dilución Immunocytochemistry/Immunofluorescence
Formulación 50mM Sodium Borate
Alias de gen FLJ12634, KIAA0518, MAD5, MAX dimerization protein 5, MAX gene associated, MAX gene-associated protein, MXD5
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with E. coli-derived Human MGA fragment.
Método de purificación Antigen Affinity-purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 23269
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.