Descripción
METTL14 Monoclonal antibody specifically detects METTL14 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | METTL14 |
| Aplicaciones | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | CL4252 |
| Conjugado | mFluor Violet 610 SE |
| Formulación | 50mM Sodium Borate |
| Alias de gen | EC 2.1.1, EC 2.1.1.-, KIAA1627methyltransferase-like protein 14, methyltransferase like 14 |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was developed using a recombinant protein derived from Q9HCE5, with the exact immunogen sequence remaining proprietary. |
| Método de purificación | Protein A purified |
| Mostrar más |