missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hidroxi-4-yodobenzoato de metilo, 97 %, Thermo Scientific™
37.47€ - 436.00€
Identificadores de productos químicos
CAS | 157942-12-6 |
---|---|
Fórmula molecular | C8H7IO3 |
Peso molecular (g/mol) | 278.045 |
Número MDL | MFCD06203960 |
Clave InChI | LXCQVWRESZDFGW-UHFFFAOYSA-N |
Sinónimo | methyl-4-iodo-3-hydroxy benzoate, 3-hydroxy-4-iodobenzoic acid methyl ester, 3-hydroxy-4-iodo-benzoic acid methyl ester, methyl 3-hydroxy-4-iodobenzenecarboxylate, benzoic acid, 3-hydroxy-4-iodo-, methyl ester, rarechem al bf 0964, methyl 3-hydroxy4-iodobenzoate, methyl 4-iodo-3-hydroxybenzoate, methyl-3-hydroxy-4-iodobenzoate, methyl-4-iodo-3-hydroxybenzoate |
PubChem CID | 10378812 |
Nombre IUPAC | 3-hidroxi-4-yodobenzoato de metilo |
SMILES | COC(=O)C1=CC(=C(C=C1)I)O |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
AplicacionesIntermedio útil en la síntesis farmacéutica y orgánica.
Solubilidad
Ligeramente soluble en agua.
Notas
Sensible a la luz. Almacenar en un lugar oscuro. Almacenar alejado de agentes oxidantes.
Identificadores de productos químicos
157942-12-6 | |
278.045 | |
LXCQVWRESZDFGW-UHFFFAOYSA-N | |
10378812 | |
COC(=O)C1=CC(=C(C=C1)I)O |
C8H7IO3 | |
MFCD06203960 | |
methyl-4-iodo-3-hydroxy benzoate, 3-hydroxy-4-iodobenzoic acid methyl ester, 3-hydroxy-4-iodo-benzoic acid methyl ester, methyl 3-hydroxy-4-iodobenzenecarboxylate, benzoic acid, 3-hydroxy-4-iodo-, methyl ester, rarechem al bf 0964, methyl 3-hydroxy4-iodobenzoate, methyl 4-iodo-3-hydroxybenzoate, methyl-3-hydroxy-4-iodobenzoate, methyl-4-iodo-3-hydroxybenzoate | |
3-hidroxi-4-yodobenzoato de metilo |
Especificaciones
Methyl 3-hydroxy-4-iodobenzoate | |
C8H7IO3 | |
7289690 | |
methyl-4-iodo-3-hydroxy benzoate, 3-hydroxy-4-iodobenzoic acid methyl ester, 3-hydroxy-4-iodo-benzoic acid methyl ester, methyl 3-hydroxy-4-iodobenzenecarboxylate, benzoic acid, 3-hydroxy-4-iodo-, methyl ester, rarechem al bf 0964, methyl 3-hydroxy4-iodobenzoate, methyl 4-iodo-3-hydroxybenzoate, methyl-3-hydroxy-4-iodobenzoate, methyl-4-iodo-3-hydroxybenzoate | |
LXCQVWRESZDFGW-UHFFFAOYSA-N | |
3-hidroxi-4-yodobenzoato de metilo | |
10378812 | |
97% |
157942-12-6 | |
MFCD06203960 | |
Light sensitive | |
Slightly soluble in water. | |
COC(=O)C1=CC(=C(C=C1)I)O | |
278.045 | |
278.05 | |
Sólido |
Seguridad y manipulación
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes
RUO – Research Use Only