Learn More
Clorhidrato de 1-aminociclobutanocarboxilato de metilo, 95 %
54.60€ - 650.00€
CAS | 92398-47-5 |
---|---|
Fórmula molecular | C6H11NO2 |
Peso molecular (g/mol) | 129.16 |
Número MDL | MFCD01318265 |
Clave InChI | SGXOPTPGZBLQAY-UHFFFAOYSA-N |
Sinónimo | methyl 1-aminocyclobutanecarboxylate, 1-amino-cyclobutanecarboxylic acid methyl ester, 1-aminocyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, 1-amino-, methyl ester, 1-amino-cyclobutanecarboxylicacidmethylester, methyl 1-amino-cyclobutane-1-carboxylate, 1-aminocyclobutane-1-carboxylic acid methyl ester, cyclobutanecarboxylicacid, 1-amino-, methyl ester |
PubChem CID | 1502033 |
Nombre IUPAC | 1-aminociclobutano-1-carboxilato de metilo |
SMILES | COC(=O)C1(N)CCC1 |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
AplicacionesSe utiliza como intermedio primario y secundario en síntesis farmacéutica y orgánica.
Solubilidad
Ligeramente soluble en agua.
Notas
Sensible al aire. Almacenar alejada del aire. Almacenar en un lugar fresco y seco en un recipiente cerrado herméticamente. Garantice una ventilación adecuada. Almacenar alejado de agentes oxidantes.
92398-47-5 | |
129.16 | |
SGXOPTPGZBLQAY-UHFFFAOYSA-N | |
1502033 | |
COC(=O)C1(N)CCC1 |
C6H11NO2 | |
MFCD01318265 | |
methyl 1-aminocyclobutanecarboxylate, 1-amino-cyclobutanecarboxylic acid methyl ester, 1-aminocyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, 1-amino-, methyl ester, 1-amino-cyclobutanecarboxylicacidmethylester, methyl 1-amino-cyclobutane-1-carboxylate, 1-aminocyclobutane-1-carboxylic acid methyl ester, cyclobutanecarboxylicacid, 1-amino-, methyl ester | |
1-aminociclobutano-1-carboxilato de metilo |
Especificaciones
Methyl 1-aminocyclobutanecarboxylate hydrochloride | |
C6H11NO2 | |
5615121 | |
methyl 1-aminocyclobutanecarboxylate, 1-amino-cyclobutanecarboxylic acid methyl ester, 1-aminocyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, 1-amino-, methyl ester, 1-amino-cyclobutanecarboxylicacidmethylester, methyl 1-amino-cyclobutane-1-carboxylate, 1-aminocyclobutane-1-carboxylic acid methyl ester, cyclobutanecarboxylicacid, 1-amino-, methyl ester | |
SGXOPTPGZBLQAY-UHFFFAOYSA-N | |
1-aminociclobutano-1-carboxilato de metilo | |
1502033 | |
95% |
92398-47-5 | |
MFCD01318265 | |
Air Sensitive | |
Slightly soluble in water. | |
COC(=O)C1(N)CCC1 | |
129.16 | |
165.62 | |
Sólido |
Seguridad y manipulación
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Temperatura ambiente; almacenar en nitrógeno.