Learn More
Methionine Aminopeptidase 2/METAP2 Antibody, Novus Biologicals™
Código de producto. 18656676 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microlitros
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18656676 100 μg 100 microlitros
18646977 25 μg 25 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18656676 Proveedor Novus Biologicals N.º de proveedor NBP305513100ul
Rabbit Polyclonal Antibody
Methionine Aminopeptidase 2/METAP2 Polyclonal antibody specifically detects Methionine Aminopeptidase 2/METAP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Methionine Aminopeptidase 2/METAP2
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulación PBS, pH 7.2, containing 40% glycerol
Alias de gen eIF-2-associated p67 homolog, Initiation factor 2-associated 67 kDa glycoprotein, MAP 2, MAP2, MetAP 2, methionine aminopeptidase 2, methionyl aminopeptidase 2, MNPEP, p67, p67EC 3.4.11.18, P67EIF2, Peptidase M 2
Especie del huésped Rabbit
Inmunógeno This antibody was developed against a recombinant protein corresponding to amino acids: SGSHLNGDLDPDDREEGAASTAEEAAKKKRRKKKKSKGPSAAGEQEPDKESGASVDEVARQLERSALEDKERDEDDEDGDGDGDGATGKKKK
Método de purificación Affinity purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 10988
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG