Learn More
Methionine Aminopeptidase 2/METAP2 Antibody, Novus Biologicals™
Código de producto. 18646977 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microlitros
25 microlitros
18646977 25 μg 25 microlitros
18656676 100 μg 100 microlitros
2 options
Código de producto. 18646977
Marca: Novus Biologicals NBP30551325ul
Rabbit Polyclonal Antibody
Methionine Aminopeptidase 2/METAP2 Polyclonal antibody specifically detects Methionine Aminopeptidase 2/METAP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Methionine Aminopeptidase 2/METAP2
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulación PBS, pH 7.2, containing 40% glycerol
Alias de gen eIF-2-associated p67 homolog, Initiation factor 2-associated 67 kDa glycoprotein, MAP 2, MAP2, MetAP 2, methionine aminopeptidase 2, methionyl aminopeptidase 2, MNPEP, p67, p67EC 3.4.11.18, P67EIF2, Peptidase M 2
Especie del huésped Rabbit
Inmunógeno This antibody was developed against a recombinant protein corresponding to amino acids: SGSHLNGDLDPDDREEGAASTAEEAAKKKRRKKKKSKGPSAAGEQEPDKESGASVDEVARQLERSALEDKERDEDDEDGDGDGDGATGKKKK
Método de purificación Affinity purified
Cantidad 25 μg
Estado normativo RUO
Disciplina de investigación Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 10988
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG