missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metapneumovirus Nucleoprotein Antibody (hMPV123), Janelia Fluor™ 549, Novus Biologicals™
Tienda Bio Techne ProductosDescription
Metapneumovirus Nucleoprotein Monoclonal antibody specifically detects Metapneumovirus Nucleoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antígeno | Metapneumovirus Nucleoprotein |
| Aplicaciones | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | hMPV123 |
| Conjugado | Janelia Fluor 549 |
| Formulación | 50 mM Sodium Borate |
| Especie del huésped | Mouse |
| Inmunógeno | Human MPV strain NCL-145 |
| Método de purificación | Protein A purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?