missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEMO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14604-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
May control cell migration by relaying extracellular chemotactic signals to the microtubule cytoskeleton. Mediator of ERBB2 signaling. The MEMO1-RHOA-DIAPH1 signaling pathway plays an important role in ERBB2-dependent stabilization of microtubules at the cell cortex. It controls the localization of APC and CLASP2 to the cell membrane, via the regulation of GSK3B activity. In turn, membrane-bound APC allows the localization of the MACF1 to the cell membrane, which is required for microtubule capture and stabilization. Is required for breast carcinoma cell migration.Especificaciones
| MEMO1 | |
| Polyclonal | |
| Unconjugated | |
| MEMO1 | |
| C21orf19 like protein, C2orf4, CGI 27, mediator of cell motility 1, MEMO, Memo 1, MEMO1, NS5ATP7, Protein MEMO1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 298787, 51072, 76890 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4QQR9, Q91VH6, Q9Y316 | |
| MEMO1 | |
| MEMO1 Fusion Protein Ag6151 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido