Learn More
B Braun™ Meliseptol™ New Formula 1000 mL Bottle
Click to view available options
Volumen (métrico):
1000 mL
Tamaño de la unidad:
1 litro
Descripción
- New – available in a perfume-free version for use in the food sector
- Optimized cleaning performance, even on shining metallic surfaces
- Without additional antimicrobial ingredient (as aldehydes, QAC)
- Fresh and pleasant smell
- Fast and comprehensive activity spectrum including Mycobacteria and non-enveloped viruses Rota-, Adeno-, Norovirus (MNV)
Especificaciones
Especificaciones
| pH | ∼7 |
| Aspecto | Incolora |
| Tipo de recipiente | Frasco |
| Para utilizar con (aplicación) | Rapid surface disinfection |
| Incluye | Without spray head |
| Volumen (métrico) | 1000 mL |