Learn More
Eurofins DiscoverX MEK1 Protein, Active
Descripción
ERKs (extracellular signal-regulated kinase) are activated by the MEK (mitogen-activated protein kinase, or ERK kinase). MEKs are activated by members of the Raf family. The signaling pathway comprising Raf and ERK (extracellular signal-regulated kinase) lies downstream of the small guanine nucleotide binding protein Ras and mediates several apparently conflicting cellular responses, such as proliferation, apoptosis, growth arrest, differentiation, and senescence, depending on the duration and strength of the external stimulus and on cell type. An inhibitor assay may be of benefit for the treatment of cancer, diabetes, neurodegenerative disorders or asthma.
EMBL Z30163, possessing amino acid substitution A327G. Please note, introduction of the A327G mutation to the rabbit sequence (Z30163) results in expression of recombinant MEK1 protein that is 100% identical to wild-type human MEK1 described in SwissProt Q02750, note this SwissProt entry fails to record M1 and starts at P2.
Especificaciones
Especificaciones
| Formulación | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Peso molecular | 71 kDa |
| Cantidad | 250 μg |
| Especie | Human |
| Recombinante | Recombinant |
| Etiqueta de proteína | N-terminal GST and C-terminal 6His tags |
| Sistema de expresión | Recombinant enzyme expressed in E. coli cells |
| Forma de proteína | Full-length |
| Grado de pureza o calidad | ≥72% |
| Expresión | MEK1, active |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.