Learn More
mCherry Antibody, Novus Biologicals™
Código de producto. 18736713 Tienda Bio Techne Productos
Change view
Click to view available options
Tamaño de la unidad:
0.10 ml
25 microlitros
Cantidad:
0.025 mL
Código de producto. unitSize Cantidad
18736713 0.10 ml -
18069453 25 microlitros 0.025 mL
2 options
Código de producto. 18736713
Marca: Novus Biologicals NBP225158
Chicken Polyclonal Antibody has been used in 26 publications
mCherry Polyclonal antibody specifically detects mCherry in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry - Free Floating, Immunohistochemistry Whole-Mount.
TRUSTED_SUSTAINABILITY
Especificaciones
Aplicaciones Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating)
Clasificación Polyclonal
Isotype IgY
For Research Use Only