missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MART-1 / MLANA Rabbit, Clone: MLANA/1761R, enQuireBio™
Rabbit Monoclonal Antibody
Marca: EnquireBio 2315-RBM11-P1
Detalles adicionales : Peso : 0.01000kg
Especificaciones
MART-1 / MLANA | |
Cytoplasmic | |
MLANA/1761R | |
Unconjugated | |
NP_005502 | |
Rabbit | |
20-22kDa (doublet) | |
Protein A/G Purified | |
RUO | |
2315 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
MLANA | |
Recombinant human full-length MLANA protein | |
SK-MEL-13 and SK-MEL-19 Melanoma cell lines; Melanomas | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |