missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MARK2, GST Tag Recombinant Protein
Click to view available options
Cantidad:
10 μg
Tamaño de la unidad:
10 microgramos
Descripción
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.
MARK2 is a serine/threonine protein kinase involved in the control of cell polarity and microtubule stability. Several cDNA clones have been isolated that encoded two isoforms of the human ser/thr protein kinase EMK1. These isoforms were characterized by the presence of a 162-bp alternative exon that gave rise to two forms, one containing the exon and the other one lacking it. Both forms were found to be coexpressed in a number of selected cell lines and tissue samples. The human EMK1 was shown to be encoded by a single mRNA ubiquitously expressed.
Especificaciones
Especificaciones
| Número de acceso | Q7KZI7 |
| Concentración | See Label |
| Para utilizar con (aplicación) | Kinase Assay |
| Formulación | 50mM tris with 50% glycerol, 2mM DTT, 0.02% Triton X-100, 0.5mM EDTA, 150mM NaCl and no preservative; pH 7.5 |
| ID de gen (Entrez) | 2011 |
| Peso molecular | 110.6 kDa |
| Nombre | Human MARK2, GST Tag |
| Intervalo de pH | 7.5 |
| Método de purificación | Purified |
| Cantidad | 10 μg |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido