missing translation for 'onlineSavingsMsg'
Learn More

MAP2 Antibody (5H11), Novus Biologicals™

Código de producto. 18747223 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mL
0.1 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18747223 0.1 mL 0.10 ml
18050094 0.025 mL 25 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18747223 Proveedor Novus Biologicals N.º de proveedor NBP192711

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody has been used in 3 publications

MAP2 Monoclonal specifically detects MAP2 in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno MAP2
Aplicaciones Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating)
Clasificación Monoclonal
Clon 5H11
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating
Formulación 50% PBS, 50% glycerol with 5mM Sodium Azide
N.º de referencia del gen P11137
Alias de gen DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2
Símbolos de los genes MAP2
Especie del huésped Mouse
Inmunógeno MAP2 Antibody (5H11) was developed against full length purified bovine protein, epitope mapped to projection domain of human sequence, amino acids 1057-1588 using MAP2 Projection Domain 3.
Peso molecular del antígeno 199 kDa
Método de purificación Protein G purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cellular Markers, Cytoskeleton Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 4133
Especificidad de la prueba Note that since the epitope for this antibody in within the projection domain found only in MAP2A and MAP2B, and so the antibody does not bind to the lower molecular weight MAP2C and MAP2D isoforms which lack this region.
Especies diana Human, Mouse, Rat, Bovine, Monkey
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG2b
Mostrar más Mostrar menos

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.