missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2 Antibody (2C4), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-50032
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MAP2 Monoclonal antibody specifically detects MAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Especificaciones
| MAP2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:5000 - 1:10000, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/ Immunofluorescence 1:5000 - 1:10000 | |
| P11137, P11137 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 2C4 | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
| MAP2 Antibody (2C4) was developed against full length recombinant human MAP2D. [UniProt# P11137] | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers | |
| 4133 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido