missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mammaglobin A Antibody (CL8892) [mFluor Violet 610 SE], Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Mammaglobin A Monoclonal antibody specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | Mammaglobin A |
| Aplicaciones | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | CL8892 |
| Conjugado | mFluor Violet 610 SE |
| Formulación | 50mM Sodium Borate |
| Alias de gen | mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1 |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was developed using a synthetic peptide derived from Q13296, with the exact immunogen sequence remaining proprietary. |
| Método de purificación | Protein A purified |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?