missing translation for 'onlineSavingsMsg'
Learn More

Maltose Binding Protein Antibody (R29), DyLight 405, Novus Biologicals™

Product Code. 18233193 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.125 mL
Unit Size:
125 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18233193 0.125 mL 125 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18233193 Supplier Novus Biologicals Supplier No. NB10066609V

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Maltose Binding Protein Monoclonal specifically detects Maltose Binding Protein in Bacteria samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Maltose Binding Protein
Aplicaciones Western Blot
Clasificación Monoclonal
Clon R29
Conjugado DyLight 405
Dilución Western Blot
Formulación 50mM Sodium Borate with 0.05% Sodium Azide
Alias de gen Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein
Símbolos de los genes malE
Especie del huésped Mouse
Inmunógeno Recombinant maltose binding protein
Método de purificación Protein G purified
Cantidad 0.125 mL
Estado normativo RUO
Disciplina de investigación Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 948538
Especificidad de la prueba Recognizes maltose binding protein (MBP). The is useful in recognizing proteins expressed with an MBP tag.
Especies diana Bacteria
Contenido y almacenamiento Store at 4C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.