missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
MAGEB4 Monoclonal antibody specifically detects MAGEB4 in Human samples. It is validated for Immunohistochemistry,Western Blot
Especificaciones
Especificaciones
| Antígeno | MAGEB4 |
| Aplicaciones | Immunohistochemistry, Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI3A3 |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:2000, Immunohistochemistry 1:500 |
| Formulación | PBS (pH 7.3) 1% BSA, 50% glycerol |
| Alias de gen | cancer/testis antigen family 3, member 6, CT3.6, MAGE-B4 antigen, melanoma antigen family B, 4, melanoma-associated antigen B4, MGC33144 |
| Especie del huésped | Mouse |
| Inmunógeno | Human recombinant protein fragment corresponding to amino acids 1-193 of human MAGEB4(NP_002358) produced in E.coli. |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?