missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
MACC1 Polyclonal antibody specifically detects MACC1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | MACC1 |
| Aplicaciones | ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | FITC |
| Formulación | PBS |
| Alias de gen | metastasis associated in colon cancer 1, SH3 domain-containing protein 7a5,7A5 |
| Especie del huésped | Rabbit |
| Inmunógeno | MACC1 antibody was raised against a 17 amino acid synthetic peptide near the center of human MACC1. The immunogen is located within amino acids 430 - 480 of MACC1. Amino Acid Squence: PDFEVKTEGERKEIKQK |
| Método de purificación | Peptide affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?