missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-108 Monoclonal Antibody (eBio13G3-19D (13G3-19D)), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
192.00€ - 360.00€
Especificaciones
| Antígeno | Ly-108 |
|---|---|
| Clon | eBio13G3-19D (13G3-19D) |
| Concentración | 0.1 mg/mL |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
Description
Ly108 is the mouse homologue of human SLAMF6 (signaling lymphocyte activation molecule family member 6). Mouse Ly108 is expressed on NK, B cells, and T cells. Activation of Ly108 receptor enhances NK cell activity and IFN-gamma expression.Specifications
| Ly-108 | |
| 0.1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Activating NK receptor; CD352; KAL1; KAL1b; KALI; KALIb; Ly108; Lymphocyte antigen 108; natural killer-, T- and B-cell antigen; NK-T-B-antigen; NTBA; NTB-A; NTBA receptor; RGD1561848; SF2000; SLAM family member 6; Slamf6; UNQ6123/PRO20080 | |
| SLAMF6 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! |
| eBio13G3-19D (13G3-19D) | |
| Flow Cytometry | |
| NovaFluor Red 700 | |
| Mouse | |
| Mouse | |
| Q9ET39 | |
| 30925 | |
| IgG2a κ | |
| Affinity chromatography | |
| SLAMF6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title