missing translation for 'onlineSavingsMsg'
Learn More

LMO2 Antibody (LMO2/3147R), DyLight 594, Novus Biologicals™

Código de producto. 18590656 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
18590656 100 μg 100 microlitros
1 options
Este artículo no se puede devolver. View return policy

Product Code. 18590656

Brand: Novus Biologicals NBP308945DL594

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

LMO2 Monoclonal antibody specifically detects LMO2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antígeno LMO2
Aplicaciones Flow Cytometry, Immunofluorescence
Clasificación Monoclonal
Clon LMO2/3147R
Conjugado DyLight 594
Formulación 50mM Sodium Borate
Alias de gen Cysteine-rich protein TTG-2, LIM domain only 2 (rhombotin-like 1), LIM domain only protein 2, LMO-2, RBTN2rhombotin-2, RBTNL1rhombotin-like 1, RHOM2T-cell translocation protein 2, T-cell translocation gene 2, TTG2rhombotin 2
Especie del huésped Rabbit
Inmunógeno Recombinant fragment (around aa 23-140) human LMO2 protein (exact sequence is proprietary) (Uniprot: P25791)
Método de purificación Protein A or G purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Angiogenesis, Cancer
Primario o secundario Primary
ID de gen (Entrez) 4005
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.