Learn More
Novus Biologicals™ Liver Matched Tumor and Normal Tissue Lysate
Shop All Bio Techne Products
Click to view available options
Cantidad:
2 viales
Unit Size:
paquete de 2
Description
The Liver Matched Tumor & Normal Tissue Lysate is derived from human.
Specifications
Specifications
| Tissue | Liver |
| Cantidad | 2 viales |
| Especie del huésped | Human |
| Contenido y almacenamiento | Store at −80°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction