missing translation for 'onlineSavingsMsg'
Learn More

LINGO-1 Antibody, Novus Biologicals™

Código de producto. 18268973 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
Código de producto. Cantidad unitSize
18268973 0.025 mL 25 microlitros
18751644 - 0.10 ml
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18268973

Marca: Novus Biologicals NBP1463350.025ML

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LINGO-1 Polyclonal specifically detects LINGO-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno LINGO-1
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Western Blot
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulación Whole antisera with No Preservative
Alias de gen FLJ14594, LERN1UNQ201, leucine rich repeat and Ig domain containing 1, leucine rich repeat neuronal 6A, Leucine-rich repeat and immunoglobilin domain-containing protein 1, Leucine-rich repeat neuronal protein 1, Leucine-rich repeat neuronal protein 6A, LRRN6Aleucine-rich repeat neuronal 6A, MGC17422
Símbolos de los genes LINGO1
Especie del huésped Rabbit
Inmunógeno A synthetic peptide fromc-terminal region of human Lingo1 conjugated to an immunogenic carrier protein was used as the antigen. The peptide is homologous in many other species.
Método de purificación Unpurified
Cantidad 0.025 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 84894
Especificidad de la prueba Specific for Lingo1.
Reconstitución Reconstitute with 0.1 ml sterilized water to desired concentration.
Especies diana Mouse, Human, Rat
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.