missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
lin-54 Polyclonal antibody specifically detects lin-54 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
Specifications
| Antígeno | lin-54 |
| Aplicaciones | Western Blot, Immunoprecipitation |
| Clasificación | Polyclonal |
| Conjugado | FITC |
| Dilución | Western Blot, Immunoprecipitation |
| Formulación | PBS |
| Alias de gen | CXCDC1, JC8.6, MIP120, Protein LIN-54 |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody maps to a region between residue 1 and 50 of human Lin-54 homolog using the numbering given in entry NP_919258.2 (GeneID 132660). |
| Método de purificación | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?