Learn More
LIN-28A Antibody, Novus Biologicals™
Código de producto. 18785233 Tienda Bio Techne Productos
Change view
Click to view available options
Tamaño de la unidad:
0.10 ml
25 microlitros
Cantidad:
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. unitSize Cantidad
18785233 25 microlitros -
18777673 0.10 ml 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18785233 Proveedor Novus Biologicals N.º de proveedor NBP149537SS
Rabbit Polyclonal Antibody has been used in 3 publications
LIN-28A Polyclonal specifically detects LIN-28A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Aplicaciones Immunohistochemistry, Immunofluorescence
Clasificación Polyclonal
Especies diana Human, Mouse
Contenido y almacenamiento Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Formulario Antisera
Isotype IgG
For Research Use Only