Descripción
Unlike standard AMCA labeling procedures, where a large molar excess of AMCA is employed, the Lightning-Link AMCA kit combines a low molar ratio of AMCA:antibody and high labeling efficiency to eliminate the need for purification steps. AMCA (aminomethylcoumarin acetate) is a blue fluorescent dye commonly used in fluorescence microscopy, arrays and in-situ hybridisation. It has an excitation wavelength of 345-350nm and an emission wavelength of 440-460nm.
Información de órdenes
Cantidad
Especificaciones
Especificaciones
| Contenido y almacenamiento | Store at -20°C. Avoid freeze-thaw cycles. |
| Cantidad | 1 mg |
| Conjugado | AMCA |
| Contenido del kit | Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent |
| Tipo de producto | Rapid AMCA Antibody Labeling Kit |